Data Logging Sound Level Meter, Model SDL600, Extech
Supplier: Avantor
Certificates
About this item
Sound Level Meter/Datalogger that records data on an SD card in Excel® format
The SDL600 features Auto or Manual ranging and a large backlit LCD display. It can store 99 readings manually and 20M readings via 2G SD card. Datalogger date/time stamps and stores readings on an SD card in Excel® format for easy transfer to a PC. Other features include Min/Max, Data Hold, Auto Power Off, and Built-in PC interface. AC analog output. A and C weighting .
Certifications: CE marked.
Ordering information: Complete with 6 AA batteries, SD Card, wind screen, and hard carrying case. Three year warranty.