Salt Refractometer, with ATC, 0 to 100%, Sper Scientific
Catalog # 66214-990
Supplier: Sper Scientific
Specifications
- Description:Salt refractometer, with ATC, 0 – 100 ppt
- Cat. No.:66214-990
- Supplier no.:300011
Specifications
About this item
Refractometer features Automatic Temperature Compensation (ATC), which makes manual temperature corrections unnecessary
Double scale in parts per thousand and specific gravity. Measures salt content in sea water, aquariums, breeding ponds, or pickling-brines. While the scale is calibrated for sodium chloride, it can also be used for calcium chloride, barium cobalt, magnesium, cesium, potassium, sodium chloride, cupric sulfate, potassium iodide, citric acid, and acetic acid (vinegar), when tested against a known standard. To operate, simply place a drop or two of sample on the prism and read the results.
Specific gravity: 1.000 to 1.070
Ordering information: Refractometer is supplied with a hard carrying case, calibration tool, and 5-year warranty.