Pig Neuropeptide Y (13-36) Trifluoroacetate
Catalog # H-9300.0001BA
Supplier: Bachem Americas
CAS Number:
Specifications
- CAS No.:113662-54-7
- Species:Pig
- Size:1 mg
- Storage Conditions:–20 ± 5 °C
- MDL No.:MFCD18782504
- Protein/Peptide Name:Neuropeptide Y (13-36) Trifluoroacetate
- Molecular Weight:2982.40
- Sequence:H-Pro-Ala-Glu-Asp-Leu-Ala-Arg-Tyr-Tyr-Ser-Ala-Leu-Arg-His-Tyr-Ile-Asn-Leu-Ile-Thr-Arg-Gln-Arg-Tyr-NH₂
- Molecular Formula:C₁₃₅H₂₀₉N₄₁O₃₆
- Cat. No.:H-9300.0001BA
- Supplier no.:H-9300.0001
Specifications
About this item
This fragment is able to compete with the intact NPY for essentially all binding sites in rat brain. However, it is less potent than the native peptide in its NPY-receptor binding affinity.