Order Entry
Puerto Rico
ContactUsLinkComponent
Rat Copeptin Trifluoroacetate
Rat Copeptin Trifluoroacetate
Supplier:  Bachem Americas
Certificates
Rat Copeptin Trifluoroacetate
Supplier:  Bachem Americas

Specifications

  • CAS No.:
    86280-64-0
  • Species:
    Rat
  • Storage Conditions:
    –20 ± 5 °C
  • Protein/Peptide Name:
    Copeptin Trifluoroacetate
  • Molecular Weight:
    4281.80
  • Sequence:
    H-Ala-Arg-Glu-Gln-Ser-Asn-Ala-Thr-Gln-Leu-Asp-Gly-Pro-Ala-Arg-Glu-Leu-Leu-Leu-Arg-Leu-Val-Gln-Leu-Ala-Gly-Thr-Gln-Glu-Ser-Val-Asp-Ser-Ala-Lys-Pro-Arg-Val-Tyr-OH
  • Molecular Formula:
    C₁₈₃H₃₀₇N₅₇O₆₁

Specifications