Mouse Urocortin II Trifluoroacetate
Supplier: Bachem Americas
Certificates
About this item
Urocortin II (mouse) exhibited considerably higher affinity for both splice variants of the type 2 CRF receptor (CRF-R2) than for CRF-R1. The potencies of Ucn II (mouse) in activating CRF-R2α and CRF-R2β were nearly equivalent to that of urocortin (rat).
Specifications
- CAS No.:330648-32-3
- Species:Mouse
- Storage Conditions:–20 ± 5 °C
- Protein Synonyms:Ucn II
- Protein/Peptide Name:Urocortin II Trifluoroacetate
- Molecular Weight:4152.95
- Sequence:H-Val-Ile-Leu-Ser-Leu-Asp-Val-Pro-Ile-Gly-Leu-Leu-Arg-Ile-Leu-Leu-Glu-Gln-Ala-Arg-Tyr-Lys-Ala-Ala-Arg-Asn-Gln-Ala-Ala-Thr-Asn-Ala-Gln-Ile-Leu-Ala-His-Val-NH₂
- Molecular Formula:C₁₈₇H₃₂₀N₅₆O₅₀