Order Entry
Puerto Rico
ContactUsLinkComponent
 
Bovine;Human;Rat (Phe²,Nle⁴)-ACTH (1-24) Trifluoroacetate
 
Bovine;Human;Rat (Phe²,Nle⁴)-ACTH (1-24) Trifluoroacetate

 

  • CAS No.:
    97773-00-7
  • Species:
    Human
    Bovine
    Rat
  • Storage Conditions:
    –20 ± 5 °C
  • MDL No.:
    MFCD18782350
  • Protein/Peptide Name:
    (Phe²,Nle⁴)-ACTH (1-24) Trifluoroacetate
  • Molecular Weight:
    2899.44
  • Sequence:
    H-Ser-Phe-Ser-Nle-Glu-His-Phe-Arg-Trp-Gly-Lys-Pro-Val-Gly-Lys-Lys-Arg-Arg-Pro-Val-Lys-Val-Tyr-Pro-OH
  • Molecular Formula:
    C₁₃₇H₂₁₂N₄₀O₃₀