Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse;Rat ACTH (1-39) Trifluoroacetate
Mouse;Rat ACTH (1-39) Trifluoroacetate
Supplier:  Bachem Americas
Certificates
Mouse;Rat ACTH (1-39) Trifluoroacetate
Supplier:  Bachem Americas

Specifications

  • CAS No.:
    77465-10-2
  • Species:
    Rat
    Mouse
  • Storage Conditions:
    –20 ± 5 °C
  • MDL No.:
    MFCD00167431
  • Protein Synonyms:
    Adrenocorticotropic hormone
  • Protein/Peptide Name:
    ACTH (1-39) Trifluoroacetate
  • Molecular Weight:
    4582.23
  • Sequence:
    H-Ser-Tyr-Ser-Met-Glu-His-Phe-Arg-Trp-Gly-Lys-Pro-Val-Gly-Lys-Lys-Arg-Arg-Pro-Val-Lys-Val-Tyr-Pro-Asn-Val-Ala-Glu-Asn-Glu-Ser-Ala-Glu-Ala-Phe-Pro-Leu-Glu-Phe-OH
  • Molecular Formula:
    C₂₁₀H₃₁₅N₅₇O₅₇S

Specifications