Bovine;Human;Rat (D-Lys¹⁶)-ACTH (1-24) Trifluoroacetate
Supplier: Bachem Americas
Certificates
About this item
Please see also α-MSH (corresponds to acetyl-ACTH (1-13) amide, H-1075), α-MSH (free acid) (acetyl-ACTH (1-13), H-1070), and (Des-acetyl)-α-MSH (H-4390). For reviews on ACTH and related peptides see: D.de Wied: Pituitary Adrenal System Hormones and Behaviour. Symposium on Developments in Endocrinology (1976) / Anon.: ACTH and Related Peptides: Structure, Regulation, and Action. Ann. N.Y. Acad. Sci. 297, 1 (1977) / A.V.Schally: Aspects of Hypothalamic Regulation of the Pituitary Gland. Science 202, 18 (1978) / R.Schwyzer: Studies on Polypeptide Receptors. A Critical View on the Mechanism of ACTH Action. Bull. Schweiz. Acad. Med. Wiss. 34, 263 (1978).
Specifications
- CAS No.:494750-52-6
- Species:HumanBovineRat
- Storage Conditions:–20 ± 5 °C
- MDL No.:MFCD03787950
- Protein Synonyms:(D-Lys16)-Tetracosactide
- Protein/Peptide Name:(D-Lys¹⁶)-ACTH (1-24) Trifluoroacetate
- Molecular Weight:2933.48
- Sequence:H-Ser-Tyr-Ser-Met-Glu-His-Phe-Arg-Trp-Gly-Lys-Pro-Val-Gly-Lys-D-Lys-Arg-Arg-Pro-Val-Lys-Val-Tyr-Pro-OH
- Molecular Formula:C₁₃₆H₂₁₀N₄₀O₃₁S