(d(CH₂)₅¹,D-Tyr(Et)²,Val⁴,Arg⁸,des-Gly⁹)-Vasopressin
Supplier: Bachem Americas
Certificates
About this item
For desmopressin see H-7675.
SKF 101926 is am antagonist of both the antidiuretic and vasopressor responses of vasopressin.
Specifications
- CAS No.:90332-82-4
- Storage Conditions:–20 ± 5 °C
- MDL No.:MFCD00076751
- Protein Synonyms:SK&F 101926|LS-186846
- Protein/Peptide Name:(d(CH₂)₅¹,D-Tyr(Et)²,Val⁴,Arg⁸,des-Gly⁹)-Vasopressin
- Molecular Weight:1079.36
- Sequence:β-Mercapto-β,β-cyclopentamethylene-propionyl-D-Tyr(Et)-Phe-Val-Asn-Cys-Pro-Arg-NH₂
- Molecular Formula:C₅₁H₇₄N₁₂O₁₀S₂
Specifications
Frequently Bought Together