Order Entry
Puerto Rico
ContactUsLinkComponent
H-Arg-Gly-Asp-Cys-OH Trifluoroacetate, Bachem
H-Arg-Gly-Asp-Cys-OH Trifluoroacetate, Bachem
Supplier:  Bachem Americas
Certificates
H-Arg-Gly-Asp-Cys-OH Trifluoroacetate, Bachem
Supplier:  Bachem Americas

Specifications

  • CAS No.:
    109292-46-8
  • Storage Conditions:
    –20 ± 5 °C
  • MDL No.:
    MFCD00076451
  • Protein Synonyms:
    RGDC
  • Protein/Peptide Name:
    H-Arg-Gly-Asp-Cys-OH Trifluoroacetate
  • Molecular Weight:
    449.49
  • Sequence:
    H-Arg-Gly-Asp-Cys-OH
  • Molecular Formula:
    C₁₅H₂₇N₇O₇S

Specifications