(N-Me-Tyr¹,N-Me-Arg⁷,D-Leu-NHEt⁸)-Dynorphin A (1-8) Trifluoroacetate
Catalog # H-3018.0500BA
Supplier: Bachem Americas
CAS Number:
Specifications
- CAS No.:103613-84-9
- Size:0.5 mg
- Storage Conditions:–20 ± 5 °C
- MDL No.:MFCD18782179
- Protein/Peptide Name:(N-Me-Tyr¹,N-Me-Arg⁷,D-Leu-NHEt⁸)-Dynorphin A (1-8) Trifluoroacetate
- Molecular Weight:1036.29
- Sequence:N-Me-Tyr-Gly-Gly-Phe-Leu-Arg-N-Me-Arg-D-Leu-NHEt
- Molecular Formula:C₅₀H₈₁N₁₅O₉
- Cat. No.:H-3018.0500BA
- Supplier no.:H-3018.0500
Specifications
About this item
The N-terminus of dynorphin, YGGFL, corresponds to Leu-enkephalin. So, dynorphin A (1-5) amide is filed as Leu-enkephalin amide (H-2745). For other opioid peptides see, the product families ‘Deltorphins and Dermorphins’, ‘Endorphins, MPF, Neoendorphins’, and ‘Opioid Peptides'.
Highly stable dynorphin-like analgesic peptide.