Cholecystokinin Octapeptide, Sulfated
About this item
Cholecystokinin (CCK) is a hormone originally isolated from porcine intestinal mucosa and described as a linear 33-amino acid peptide containing a sulfated tyrosine, which is essential for its biological activity. It has been found in mammals in both the digestive tract and the central nervous system. Among its multiple biological functions, this hormone stimulates pancreatic exocrine secretion, gallbladder contraction, and intestine motility and may also act as a neurotransmitter/neuromodulator in the central nervous system. For CCK-4 see H-3110.
CCK-8 exhibits various gastrointestinal effects as contraction of the gallbladder and stimulation of pancreatic secretion and gastrointestinal transit. In rats, CCK-8 facilitated the uptake of leptin from peripheral circulation to cerebrospinal fluid and its access to the hypothalamus. For CCK-4 see H-3110.
2 Options Available Below
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Cholecystokinin (CCK) is a hormone originally isolated from porcine intestinal mucosa and described as a linear 33-amino acid peptide containing a sulfated tyrosine, which is essential for its biological activity. It has been found in mammals in both the digestive tract and the central nervous system. Among its multiple biological functions, this hormone stimulates pancreatic exocrine secretion, gallbladder contraction, and intestine motility and may also act as a neurotransmitter/neuromodulator in the central nervous system. For CCK-4 see H-3110.
CCK-8 exhibits various gastrointestinal effects as contraction of the gallbladder and stimulation of pancreatic secretion and gastrointestinal transit. In rats, CCK-8 facilitated the uptake of leptin from peripheral circulation to cerebrospinal fluid and its access to the hypothalamus. For CCK-4 see H-3110.
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- H-2080.0001BA
- H-2080.0005BA
- Protein/Peptide Name:
- Cholecystokinin Octapeptide, sulfated
- Cholecystokinin Octapeptide, sulfated
- Protein Synonyms:
- Cholecystokinin (26-33) (sulfated)|Sincalide|CCK-8 (sulfated)
- Cholecystokinin (26-33) (sulfated)|Sincalide|CCK-8 (sulfated)
- CAS No.:
- 25126-32-3
- 25126-32-3
- MDL No.:
- MFCD00467179
- MFCD00467179
- Sequence:
- H-Asp-Tyr(SO₃H)-Met-Gly-Trp-Met-Asp-Phe-NH₂
- H-Asp-Tyr(SO₃H)-Met-Gly-Trp-Met-Asp-Phe-NH₂
- Molecular Weight:
- 1143.29
- 1143.29
- Molecular Formula:
- C₄₉H₆₂N₁₀O₁₆S₃
- C₄₉H₆₂N₁₀O₁₆S₃
- Storage Conditions:
- –20 ± 5 °C
- –20 ± 5 °C
Specifications
Product Family Options
Product Information
- SizeAvailabilityPrice
- Supplier Number: 4033010.0001Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.H-2080.0001BAProtein/Peptide NameCholecystokinin Octapeptide, sulfatedProtein SynonymsCholecystokinin (26-33) (sulfated)|Sincalide|CCK-8 (sulfated)CAS No.25126-32-3MDL No.MFCD00467179SequenceH-Asp-Tyr(SO₃H)-Met-Gly-Trp-Met-Asp-Phe-NH₂Molecular Weight1143.29Molecular FormulaC₄₉H₆₂N₁₀O₁₆S₃Storage Conditions–20 ± 5 °C - Supplier Number: 4033010.0005Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.H-2080.0005BAProtein/Peptide NameCholecystokinin Octapeptide, sulfatedProtein SynonymsCholecystokinin (26-33) (sulfated)|Sincalide|CCK-8 (sulfated)CAS No.25126-32-3MDL No.MFCD00467179SequenceH-Asp-Tyr(SO₃H)-Met-Gly-Trp-Met-Asp-Phe-NH₂Molecular Weight1143.29Molecular FormulaC₄₉H₆₂N₁₀O₁₆S₃Storage Conditions–20 ± 5 °C
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



