Order Entry
United States
ContactUsLinkComponent
 
Neurokinin B Trifluoroacetate
 
Neurokinin B Trifluoroacetate

 

  • CAS No.:
    86933-75-7
  • Storage Conditions:
    –20 ± 5 °C
  • MDL No.:
    MFCD18782128
  • Protein Synonyms:
    NKB
  • Protein/Peptide Name:
    Neurokinin B Trifluoroacetate
  • Molecular Weight:
    1210.44
  • Sequence:
    H-Asp-Met-His-Asp-Phe-Phe-Val-Gly-Leu-Met-NH₂
  • Molecular Formula:
    C₅₅H₇₉N₁₃O₁₄S₂