Order Entry
Puerto Rico
ContactUsLinkComponent
 
C-Reactive Protein (174-185)
 
C-Reactive Protein (174-185)

 

  • CAS No.:
    160369-86-8
  • Storage Conditions:
    –20 ± 5 °C
  • MDL No.:
    MFCD00236934
  • Protein Synonyms:
    CRP
  • Protein/Peptide Name:
    C-Reactive Protein (174-185)
  • Molecular Weight:
    1276.50
  • Sequence:
    H-Ile-Tyr-Leu-Gly-Gly-Pro-Phe-Ser-Pro-Asn-Val-Leu-OH
  • Molecular Formula:
    C₆₂H₉₃N₁₃O₁₆