Recombinant HIV-1 Protease (from E. coli)
Catalog # H-1256.0100BA
Supplier: Bachem Americas
CAS Number:
Specifications
- Enzyme type:Recombinant
- Source:E. coli
- Size:0.1 mg
- Storage Conditions:<−70 °C
- Enzyme Name:HIV-1 Protease
- Purity:Affinity-purified
- Cat. No.:H-1256.0100BA
- Supplier no.:H-1256.0100
Specifications
About this item
Sequence:
Crystallizable grade material. It is suitable for co-complexing with inhibitor screens.