Order Entry
Puerto Rico
ContactUsLinkComponent
ACTH (11-24)
ACTH (11-24)
Supplier:  Bachem Americas
Certificates
ACTH (11-24)
Supplier:  Bachem Americas

Specifications

  • CAS No.:
    4237-93-8
  • Storage Conditions:
    –20 ± 5 °C
  • MDL No.:
    MFCD00076175
  • Protein Synonyms:
    Adrenocorticotropic hormone
  • Protein/Peptide Name:
    ACTH (11-24)
  • Molecular Weight:
    1652.06
  • Sequence:
    H-Lys-Pro-Val-Gly-Lys-Lys-Arg-Arg-Pro-Val-Lys-Val-Tyr-Pro-OH
  • Molecular Formula:
    C₇₇H₁₃₄N₂₄O₁₆

Specifications