Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
H-Pro-Phe-OH

H-Pro-Phe-OH

Supplier:   Bachem Americas
H-Pro-Phe-OH
Supplier:  Bachem Americas
Certificates

2 Options Available Below

  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

See also E-2410, Suc-Pro-OH, the product familiy 'Antihypertensive Peptides' and the subfamily 'Bradykinin-Potentiating Peptides (BPP)'.

Substrate for human kidney prolinase (prolyl dipeptidase). ACE2 inhibitor, IC₅₀ 0.15 mM.

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • G-3050.0001BA
  • G-3050.0005BA
  • Protein/Peptide Name:
  • H-Pro-Phe-OH
  • H-Pro-Phe-OH
  • CAS No.:
  • 13589-02-1
  • 13589-02-1
  • MDL No.:
  • MFCD00037869
  • MFCD00037869
  • Sequence:
  • H-Pro-Phe-OH
  • H-Pro-Phe-OH
  • Molecular Weight:
  • 262.31
  • 262.31
  • Molecular Formula:
  • C₁₄H₁₈N₂O₃
  • C₁₄H₁₈N₂O₃
  • Storage Conditions:
  • –20 ± 5 °C
  • –20 ± 5 °C

Specifications

Product Family Options

Product Information

  • Size
    Availability
    Price
  • 1 g
    Log in for availability
    Log in for availability
    Each (1g)
    Each (1g) - $338.24
    $338.24
    Each (1g)
       $338.24
    Minimum order quantity is 1

    Supplier Number: 4001974.0001
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    G-3050.0001BA
    Protein/Peptide Name
    H-Pro-Phe-OH
    CAS No.
    13589-02-1
    MDL No.
    MFCD00037869
    Sequence
    H-Pro-Phe-OH
    Molecular Weight
    262.31
    Molecular Formula
    C₁₄H₁₈N₂O₃
    Storage Conditions
    –20 ± 5 °C
  • 5 g
    Log in for availability
    Log in for availability
    Each (5g)
    Each (5g) - $1,352.93
    $1,352.93
    Each (5g)
       $1,352.93
    Minimum order quantity is 1

    Supplier Number: 4001974.0005
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    G-3050.0005BA
    Protein/Peptide Name
    H-Pro-Phe-OH
    CAS No.
    13589-02-1
    MDL No.
    MFCD00037869
    Sequence
    H-Pro-Phe-OH
    Molecular Weight
    262.31
    Molecular Formula
    C₁₄H₁₈N₂O₃
    Storage Conditions
    –20 ± 5 °C