Order Entry
Puerto Rico
ContactUsLinkComponent
Chicken;Human;Mouse;Pig;Rat;Sheep PACAP-38 (28-38) Trifluoroacetate
Chicken;Human;Mouse;Pig;Rat;Sheep PACAP-38 (28-38) Trifluoroacetate
Supplier:  Bachem Americas
Certificates
Chicken;Human;Mouse;Pig;Rat;Sheep PACAP-38 (28-38) Trifluoroacetate
Supplier:  Bachem Americas

Specifications

  • CAS No.:
    160489-86-1
  • Species:
    Pig
    Sheep
    Chicken
    Human
    Rat
    Mouse
  • Storage Conditions:
    –20 ± 5 °C
  • MDL No.:
    MFCD18782302
  • Protein Synonyms:
    Pituitary Adenylate Cyclase Activating Polypeptide-38 (28-38)
  • Protein/Peptide Name:
    PACAP-38 (28-38) Trifluoroacetate
  • Molecular Weight:
    1403.70
  • Sequence:
    H-Gly-Lys-Arg-Tyr-Lys-Gln-Arg-Val-Lys-Asn-Lys-NH₂
  • Molecular Formula:
    C₆₁H₁₁₀N₂₄O₁₄

Specifications

Frequently Bought Together