Distillation Receiver, Calibrated to Contain, Ace Glass
Catalog # 89074-448
Supplier: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- Capacity:10 mL
- Description:Distillation Receiver
- Cat. No.:89074-448
- Top [ST] joint:[ST] 14/20, outer
- Subdivisions:0.2 mL
- Supplier no.:9396-02
Specifications
About this item
This distillation receiver is calibrated to contain
Fabricated from borosilicate glass.