FerroVer® Iron Reagent Powder Pillows, 10 ml, Hach
Catalog # 97026-466
Supplier: Avantor
Product available on GSA Advantage®
CAS Number:
Specifications
- Chemical method:FerroVer®
- Description:FerroVer® Iron Reagent Powder Pillows
- Size:1000 pillows
- Method:8008 - FerroVer
- Storage Temperature:10 °C to 25 °C
- Compliance:EPA - Yes
- Instrument:All except DR/3000, DR/2000
- Cat. No.:97026-466
- Range:0.02 - 3.00 mg/l Fe
- Sample Size:10 ml
- Supplier no.:2105728
Specifications
About this item
For determination of Iron by the 1, 10 phenanthroline method
USEPA approved method for reporting wastewater analysis using Hach Method 8008