Order Entry
Puerto Rico
ContactUsLinkComponent
Human Calcitonin Gene Related Peptide
Human Calcitonin Gene Related Peptide
Certificates
Human Calcitonin Gene Related Peptide
Certificates

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • CAS No.:
    90954-53-3
  • Species:
    Human
  • Storage Conditions:
    -30 °C to -10 °C
  • Form:
    Solid
  • MDL No.:
    MFCD00285485
  • Protein/Peptide Name:
    Calcitonin Gene Related Peptide
  • Solubility:
    Soluble in water or 1% acetic acid at 1mg/ml.
  • Molecular Weight:
    3789.3
  • Color:
    White to off-white
  • Molecular Formula:
    C₁₆₃H₂₆₇N₅₁O₄₉S₂

Specifications

Related Information