Order Entry
Puerto Rico
ContactUsLinkComponent
RNase Removal Reagent, Thermo Scientific
RNase Removal Reagent, Thermo Scientific
Catalog # AAJ61574-REX
CAS Number:  
RNase Removal Reagent, Thermo Scientific
Catalog # AAJ61574-REX
Supplier Number:  J61574-REX
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    475 mL
  • Solubility:
    Miscible with water
  • Density:
    1.00 g/mL
  • Physical Form:
    Liquid
  • Appearance:
    Colorless
  • Cat. No.:
    AAJ61574-REX
  • Supplier no.:
    J61574-REX

Specifications

About this item

Destroys RNases and DNases on contact.

RNase Removal Reagent is used to remove the RNase from glasswares, plastic surfaces, equipment, gel boxes, benchtops, and labware. It is also used to eliminate RNase from microcentrifuge tubes without any side enzymatic reactions.

Destroys RNases and DNases on contact