Order Entry
United States
ContactUsLinkComponent
Corning® Yeast Extract, Powder, Corning
Corning® Yeast Extract, Powder, Corning
Catalog # 45000-498
Supplier:  Corning
CAS Number:  
Corning® Yeast Extract, Powder, Corning
Catalog # 45000-498
Supplier:  Corning
Supplier Number:  61-231-RO
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Yeast Extract, Powder 10,000 MW cut-off
  • Environmentally Preferable:
  • Storage Temperature:
    Ambient
  • Shipping Temperature:
    Ambient
  • Cat. No.:
    45000-498
  • Volume:
    500 g
  • Supplier no.:
    61-231-RO

Specifications

About this item

Yeast extract is a mixture of amino acids, peptides, water soluble vitamins and carbohydrates obtained from autolyzed yeast

This is the water soluble portion with intact B-complex vitamins and can be used as an additive for culture media.