ContactUsLinkComponent
RF15 Portable Brix Refractometer with Automatic Temperature Compensation, Extech®
RF15 Portable Brix Refractometer with Automatic Temperature Compensation, Extech®
  89370-224
 :  Avantor
undefined
RF15 Portable Brix Refractometer with Automatic Temperature Compensation, Extech®
  89370-224
 :  Avantor
  • Cat. No.:
    89370-224
  • Supplier no.:
    RF15
Portable Sucrose Brix Refractometer (0–32%) with ATC that measures the concentration of sugar in fruit juice and industrial fluids
The RF15 provides accurate and repeatable measurements on an easy-to-read scale. Requires only 2 or 3 drops of solution. It features Automatic Temperature Compensation (ATC) from 10 to 30°C. Compact size, easy to operate.
 : CE marked.
 : Supplied complete with case, calibration screwdriver, and calibration solution. Refractometer also includes manufacturer's one-year warranty.