Corning® Graces's Insect Basal Medium (Vaughn Mod.), Corning
Catalog # 45000-476
Supplier: Corning
CAS Number:
Specifications
- Media Format:Liquid
- Description:Graces's Insect Basal Medium (Vaughn Mod.) with L-glutamine, without insect hemolymph
- Storage Temperature:Refrigerate
- Cat. No.:45000-476
- Supplier no.:13-200-CV
Specifications
About this item
The insect cell culture medium was formulated to closely resemble insect hemolymph, a compound crucial to the function of the insect circulatory system
The basal medium, when properly supplemented, has been used to culture cells derived from a variety of insects including several species of lepidopterans as well as some dipterans. The medium is primarily used as a basal medium for the growth and maintenance of cell lines derived from lepidopterans.