Double Stack Paper Towel Holder, PETG, TrippNT
Catalog # 89233-146
Supplier: TrippNT
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
- Cat. No.:89233-146
- W×D×H:28 x 10 x 33 cm (11 x 4 x 13")
- Weight:1 kg (2 lbs.)
- Supplier no.:50309
Specifications
About this item
Clear PETG double stack paper towel holder has a simple design that allows quick access to one or several C-fold, Z-fold, Tri-fold or multi-fold paper towels
Made from extra-tough PETG which won't break like acrylic and is clear for easy viewing. Holds about 300 folded paper towels and can sit on the counter or be mounted on the wall with included tape or keyholes.