Order Entry
Puerto Rico
ContactUsLinkComponent
Clamp, Two-Prong, Ace Glass
Clamp, Two-Prong, Ace Glass
Catalog # 89083-208
Supplier:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
undefined
Clamp, Two-Prong, Ace Glass
Catalog # 89083-208
Supplier:  Ace Glass
Supplier Number:  11075-21
Product available on GSA Advantage®
Small Business Enterprise

Specifications

  • Description:
    Two-Prong Vinylized Jaws Clamp
  • Cat. No.:
    89083-208
  • Supplier no.:
    11075-21

Specifications

About this item

These medium size clamps are have two-pronged jaws designed to holding burets up to 38mm in diameter

The position of the jaws and holder are fixed. This insures that any object held in the jaws will remain permanently in a vertical position. Clamps to rods up to 19mm.