Carbonate-Buffered Saline (5X), pH 9.5, Thermo Scientific
Supplier: Thermo Scientific Chemicals
Certificates
About this item
Liquid
CAS No.: 497-19-8
Physical Form: Liquid
MDL No.: MFCD00003494
Specifications
- CAS No.:497-19-8
- MDL No.:MFCD00003494
- Physical Form:Liquid