TEA, 0.2 M Buffer Solution, pH 8.0, Thermo Scientific
Catalog # AAJ63793-AK
Supplier: Thermo Scientific Chemicals
CAS Number:
Specifications
- CAS No.:102-71-6
- Size:250 mL
- MDL No.:MFCD00002855
- Physical Form:Liquid
- Odor/Scent:Odorless
- Appearance:Colorless
- Cat. No.:AAJ63793-AK
- Supplier no.:J63793-AK
Specifications
About this item
Liquid
CAS No.: 102-71-6
Physical Form: Liquid
Appearance: Colorless
Odor: Odorless
MDL No.: MFCD00002855