RIPA buffer (2X), Thermo Scientific
Supplier: Thermo Scientific Chemicals
About this item
Liquid
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Appearance: Colorless
MDL No.: MFCD02100484
2 Options Available Below
Product Details & Documents
Liquid
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Appearance: Colorless
MDL No.: MFCD02100484
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- AAJ63324-EQE
- AAJ63324-AK
- MDL No.:
- MFCD02100484
- MFCD02100484
- Appearance:
- Colorless
- Colorless
- Physical Form:
- Liquid
- Liquid
- Storage Temperature:
- 2°C to 8°C
- 2°C to 8°C
Specifications
Product Family Options
Product Information
- SizeAvailabilityPrice
- Supplier Number: J63324-EQESpecifications:
More
Specifications
Cat. No.AAJ63324-EQEMDL No.MFCD02100484AppearanceColorlessPhysical FormLiquidStorage Temperature2°C to 8°C - Supplier Number: J63324-AKSpecifications:
More
Specifications
Cat. No.AAJ63324-AKMDL No.MFCD02100484AppearanceColorlessPhysical FormLiquidStorage Temperature2°C to 8°C



