RIPA buffer, Thermo Scientific
Supplier: Thermo Scientific Chemicals
About this item
Liquid
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Appearance: Colorless
MDL No.: MFCD02100484
2 Options Available Below
Product Details & Documents
Liquid
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Appearance: Colorless
MDL No.: MFCD02100484
Documents
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- AAJ63306-AK
- AAJ63306-AP
- MDL No.:
- MFCD02100484
- MFCD02100484
- Appearance:
- Colorless
- Colorless
- Physical Form:
- Liquid
- Liquid
- Storage Temperature:
- 2°C to 8°C
- 2°C to 8°C
Specifications
Product Family Options
Product Information
- SizeAvailabilityPrice
- Supplier Number: J63306-AKSpecifications:
More
Specifications
Cat. No.AAJ63306-AKMDL No.MFCD02100484AppearanceColorlessPhysical FormLiquidStorage Temperature2°C to 8°C - Supplier Number: J63306-APSpecifications:
More
Specifications
Cat. No.AAJ63306-APMDL No.MFCD02100484AppearanceColorlessPhysical FormLiquidStorage Temperature2°C to 8°C
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



