Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Description:BCA Protein Assay Kit
- Size:1 kit
- Cat. No.:AAJ63283-QA
- Storage temperature:RT
- Supplier no.:J63283-QA
Specifications
About this item
BCA Protein Assay Kit can be used to measure very dilute protein concentrations in very small sample volumes.
- Contains two separate solutions for estimation of protein concentration
- Form: Liquid
- Storage: Room temperature
Accurately measures protein concentrations from 0.5 to 30 µg/ml in tube assays and 1 to 20 µg/ml in 96- or 384-well plate assays. It is used for studying protein, that is protein interactions, measuring column fractions after affinity chromatography, estimating percent recovery of membrane proteins from cell extracts and high-throughput screening of fusion proteins.