Order Entry
Puerto Rico
ContactUsLinkComponent
Saponin Permeating Solution, Thermo Scientific
Saponin Permeating Solution, Thermo Scientific
Catalog # AAJ63209-AK
CAS Number:  
Saponin Permeating Solution, Thermo Scientific
Catalog # AAJ63209-AK
Supplier Number:  J63209-AK
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    Saponin Permeating Solution
  • Size:
    250 mL
  • Storage Conditions:
    RT
  • Form:
    Liquid
  • Solubility:
    Difficult to Mix
  • Cat. No.:
    AAJ63209-AK
  • Supplier no.:
    J63209-AK

Specifications

About this item

Saponin permeating solution, is used as a cell permeabilizing agent.

Saponin permeating solution is made by mixing and dissolving 5 g of saponin with 100 ml of 5X PBS (pH 7.4) followed by submicron filtration and/or autoclaving at 15 PSI for 60 minutes.