Sodium Citrate, 0.5 M Buffer Solution, pH 5.5, Thermo Scientific
Liquid
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Appearance: Colorless
Odor: Odorless
MDL No.: MFCD00012462
- MDL No.:MFCD00012462
- Storage Temperature:2°C to 8°C
- Physical Form:Liquid
- Odor/Scent:Odorless
- Appearance:Colorless