Order Entry
Puerto Rico
ContactUsLinkComponent
Buffer Kit for Antibody Purification on Protein A Column, Thermo Scientific
Buffer Kit for Antibody Purification on Protein A Column, Thermo Scientific
Catalog # AAJ63014-QA
CAS Number:  
Buffer Kit for Antibody Purification on Protein A Column, Thermo Scientific
Catalog # AAJ63014-QA
Supplier Number:  J63014-QA
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    1kit
  • Physical Form:
    Liquid
  • Cat. No.:
    AAJ63014-QA
  • Supplier no.:
    J63014-QA

Specifications

About this item

Liquid Buffers to be used when performing antibody

Physical Form: Liquid

Applications: Buffers to be used when performing antibody purification using immobilized Protein A.

Delivery: This kit contains: 1 L of 1 M Tris-HCl (pH 8.0) and 500 mL of 100 mM Glycine (pH 3.0).