BSA Blocking Buffer, 5% in TBS, Thermo Scientific
About this item
Liquid
Storage Temperature: -30°C to -10°C
Physical Form: Liquid
2 Options Available Below
Product Details & Documents
Liquid
Storage Temperature: -30°C to -10°C
Physical Form: Liquid
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- AAJ62637-AK
- AAJ62637-AP
- Physical Form:
- Liquid
- Liquid
- Storage Temperature:
- -30°C to -10°C
- -30°C to -10°C
Specifications
Product Family Options
Product Information
- SizeAvailabilityPrice
- Supplier Number: J62637-AKSpecifications:
More
Specifications
Cat. No.AAJ62637-AKPhysical FormLiquidStorage Temperature-30°C to -10°C - Supplier Number: J62637-APSpecifications:
More
Specifications
Cat. No.AAJ62637-APPhysical FormLiquidStorage Temperature-30°C to -10°C
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



