Glycine, 0.2 M Buffer Solution, pH 3.0, Thermo Scientific
Supplier: Thermo Scientific Chemicals
Certificates
About this item
Liquid
CAS No.: 56-40-6
Physical Form: Liquid
Odor: Odorless
MDL No.: MFCD00008131
Specifications
- CAS No.:56-40-6
- MDL No.:MFCD00008131
- Physical Form:Liquid
- Odor/Scent:Odorless