Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
RIPA buffer (5X), Thermo Scientific

RIPA buffer (5X), Thermo Scientific

RIPA buffer (5X), Thermo Scientific
Certificates

2 Options Available Below

Product Details & Documents

Liquid

Storage Temperature: 2°C to 8°C
Physical Form: Liquid
MDL No.: MFCD02100484

Documents

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • AAJ62524-AD
  • AAJ62524-AE
  • MDL No.:
  • MFCD02100484
  • MFCD02100484
  • Physical Form:
  • Liquid
  • Liquid
  • Storage Temperature:
  • 2°C to 8°C
  • 2°C to 8°C

Specifications

Product Family Options

Product Information

  • Size
    Availability
    Price
  • 50 mL
    Log in for availability
    Log in for availability
    Each (50ml)
    Each (50ml) - $197.01
    $197.01
    Each (50ml)
       $197.01
    Minimum order quantity is 1

    Supplier Number: J62524-AD
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    AAJ62524-AD
    MDL No.
    MFCD02100484
    Physical Form
    Liquid
    Storage Temperature
    2°C to 8°C
  • 100 mL
    Log in for availability
    Log in for availability
    Each (100ml)
    Each (100ml) - $336.04
    $336.04
    Each (100ml)
       $336.04
    Minimum order quantity is 1

    Supplier Number: J62524-AE
    Specifications:
    More
    Specifications
    Cat. No.
    AAJ62524-AE
    MDL No.
    MFCD02100484
    Physical Form
    Liquid
    Storage Temperature
    2°C to 8°C
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.