Sodium Bicarbonate, 1 M Buffer Solution, pH 8.0, Thermo Scientific
Catalog # AAJ62495-AP
Supplier: Thermo Scientific Chemicals
CAS Number:
Specifications
- Size:500 mL
- MDL No.:MFCD00003528
- Physical Form:Liquid
- Cat. No.:AAJ62495-AP
- Supplier no.:J62495-AP
Specifications
About this item
Liquid
CAS No.: 144-55-8
Physical Form: Liquid
MDL No.: MFCD00003528