Order Entry
Puerto Rico
ContactUsLinkComponent
Blocking Reagents, BLOTTO and Tween 20 in PBS, with 0.02% Sodium Azide, Thermo Scientific
Blocking Reagents, BLOTTO and Tween 20 in PBS, with 0.02% Sodium Azide, Thermo Scientific
  AAJ62304-AK
 :  
Blocking Reagents, BLOTTO and Tween 20 in PBS, with 0.02% Sodium Azide, Thermo Scientific
  AAJ62304-AK
 :  J62304-AK
 :  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Description:
    BLOTTO and Tween 20 in PBS, with 0.02% Sodium Azide
  • Size:
    250 mL
  • Storage Temperature:
    –20 °C
  • Physical Form:
    Liquid
  • Cat. No.:
    AAJ62304-AK
  • Supplier no.:
    J62304-AK

 

 

Blocking reagent for immunohistochemistry experiments.

Contains 5% non-fat dry milk, 0.05% Tween-20 and 0.02% sodium azide in PBS.