Imidazole, 0.5 M Buffer Solution, pH 6.5, Thermo Scientific
Supplier: Thermo Scientific Chemicals
Certificates
About this item
Liquid
CAS No.: 288-32-4
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
MDL No.: MFCD00005183
Specifications
- CAS No.:288-32-4
- MDL No.:MFCD00005183
- Storage Temperature:2°C to 8°C
- Physical Form:Liquid