Order Entry
Puerto Rico
ContactUsLinkComponent
AMP, 0.2 M Buffer Solutions, Thermo Scientific
AMP, 0.2 M Buffer Solutions, Thermo Scientific
Certificates
AMP, 0.2 M Buffer Solutions, Thermo Scientific
Certificates

Specifications

  • CAS No.:
    124-68-5
  • MDL No.:
    MFCD00008051
  • Storage Temperature:
    2°C to 8°C
  • Physical Form:
    Liquid
  • Odor/Scent:
    Odorless
  • Appearance:
    Colorless

Specifications

Related Information