Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Sodium Citrate, 0.5 M Buffer Solution, pH 6.0, Thermo Scientific

Sodium Citrate, 0.5 M Buffer Solution, pH 6.0, Thermo Scientific

Sodium Citrate, 0.5 M Buffer Solution, pH 6.0, Thermo Scientific
Certificates
Sodium Citrate, 0.5 M Buffer Solution, pH 6.0, Thermo Scientific

2 Options Available Below

Product Details & Documents

Liquid

Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Appearance: Colorless
Odor: Odorless
MDL No.: MFCD00012462

Documents

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • AAJ61815-AK
  • AAJ61815-AP
  • Odor/Scent:
  • Odorless
  • Odorless
  • Physical Form:
  • Liquid
  • Liquid
  • MDL No.:
  • MFCD00012462
  • MFCD00012462
  • Appearance:
  • Colorless
  • Colorless
  • Storage Temperature:
  • 2°C to 8°C
  • 2°C to 8°C

Specifications

Product Family Options

Product Information

  • Size
    Availability
    Price
  • 250 mL
    Log in for availability
    Log in for availability
    Each (250ml)
    Each (250ml) - $159.51
    $159.51
    Each (250ml)
       $159.51
    Minimum order quantity is 1

    Supplier Number: J61815-AK
    Specifications:
    More
    Specifications
    Cat. No.
    AAJ61815-AK
    Odor/Scent
    Odorless
    Physical Form
    Liquid
    MDL No.
    MFCD00012462
    Appearance
    Colorless
    Storage Temperature
    2°C to 8°C
  • 500 mL
    Log in for availability
    Log in for availability
    Each (500ml)
    Each (500ml) - $234.43
    $234.43
    Each (500ml)
       $234.43
    Minimum order quantity is 1

    Supplier Number: J61815-AP
    Specifications:
    More
    Specifications
    Cat. No.
    AAJ61815-AP
    Odor/Scent
    Odorless
    Physical Form
    Liquid
    MDL No.
    MFCD00012462
    Appearance
    Colorless
    Storage Temperature
    2°C to 8°C
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.