Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
SM Buffer, pH 7.5, Thermo Scientific

SM Buffer, pH 7.5, Thermo Scientific

SM Buffer, pH 7.5, Thermo Scientific
Certificates

2 Options Available Below

Product Details & Documents

Liquid

Physical Form: Liquid
Applications: Phage diluent and storage buffer. Used for routine manipulation of phage suspensions.

Documents

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • AAJ61367-AE
  • AAJ61367-AK
  • Physical Form:
  • Liquid
  • Liquid

Specifications

Product Family Options

Product Information

  • Size
    Availability
    Price
  • 100 mL
    Log in for availability
    Log in for availability
    Each (100ml)
    Each (100ml) - $163.92
    $163.92
    Each (100ml)
       $163.92
    Minimum order quantity is 1

    Supplier Number: J61367-AE
    Specifications:
    More
    Specifications
    Cat. No.
    AAJ61367-AE
    Physical Form
    Liquid
  • 250 mL
    Log in for availability
    Log in for availability
    Each (250ml)
    Each (250ml) - $267.40
    $267.40
    Each (250ml)
       $267.40
    Minimum order quantity is 1

    Supplier Number: J61367-AK
    Specifications:
    More
    Specifications
    Cat. No.
    AAJ61367-AK
    Physical Form
    Liquid