TES, 0.2 M Buffer Solution, pH 7.5, Thermo Scientific
Supplier: Thermo Scientific Chemicals
Certificates
About this item
Liquid
CAS No.: 70331-82-7
Physical Form: Liquid
MDL No.: MFCD00065482
Specifications
- CAS No.:70331-82-7
- MDL No.:MFCD00065482
- Physical Form:Liquid