Lincomycin hydrochloride
About this item
Lincosamide anitbiotic active against Gram-positive bacteria
Crystalline powder
Applications: Lincosamide anitbiotic active against Gram-positive bacteria
- CAS Number:859-18-7
- Formula:C₁₈H₃₄N₂O₆S·HCl
- MDL Number:MFCD00058237
- Molecular Weight:443.0 g/mol (anhy)
- Storage Temperature:Ambient
- EINECS:212-726-7
3 Options Available Below
Product Details & Documents
Lincosamide anitbiotic active against Gram-positive bacteria
Crystalline powder
Applications: Lincosamide anitbiotic active against Gram-positive bacteria
- CAS Number:859-18-7
- Formula:C₁₈H₃₄N₂O₆S·HCl
- MDL Number:MFCD00058237
- Molecular Weight:443.0 g/mol (anhy)
- Storage Temperature:Ambient
- EINECS:212-726-7
Documents
Certificates
Search by Lot NumberAlfa Aesar_Buy 3 Get 2 Complimentary items
Promotion Material
Organic Building Blocks and Intermediates
Application Notes
Product Family Options
Product Information
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



