Sodium phosphate, 0.5 M buffer soln., pH 8.5, Thermo Scientific
Supplier: Thermo Scientific Chemicals
Certificates
About this item
Liquid
Physical Form: Liquid
MDL No.: MFCD00146206
Specifications
- MDL No.:MFCD00146206
- Physical Form:Liquid
Liquid
Physical Form: Liquid
MDL No.: MFCD00146206