Order Entry
Puerto Rico
ContactUsLinkComponent
Bovine Native Transferrin (Holo) (from Plasma)
Bovine Native Transferrin (Holo) (from Plasma)
Catalog # AAJ61046-03
CAS Number:  
Bovine Native Transferrin (Holo) (from Plasma)
Catalog # AAJ61046-03
Supplier Number:  J61046-03
CAS Number:  

Specifications

  • CAS No.:
    11096-37-0
  • Protein/Peptide Type:
    Native
  • Source:
    Plasma
  • Species:
    Bovine
  • Size:
    1 g
  • Storage Conditions:
    Refrigerator
  • Form:
    Lyophilized Powder
  • MDL No.:
    MFCD00131337
  • Protein Synonyms:
    Iron-saturated HTF bovine native protein|Siderophilin
  • Protein/Peptide Name:
    Transferrin (Holo)
  • Purity:
    >98%
  • Solubility:
    Soluble in Water (50 mg/ml)
  • Cat. No.:
    AAJ61046-03
  • Supplier no.:
    J61046-03

Specifications

About this item

Transferrin (Holo) is the primary iron binding and cell delivery molecule present in serum.

Serum-free cell culture systems require a delivery format for iron. It is the preferential delivery form of iron because cells process transferrin bound iron in a physiologically appropriate way through transferrin receptors on the cell surface. It can be used with a wide range of cells types. It is loaded with iron prior to use and can be added directly to iron poor or iron-free cell culture media. It has been used for study in treatment of breast cancer cells with holo-transferrin in combination with dihydroartemisinin results in increased tumor cell death due to elevated levels of intracellular ferrous iron.