DNA (Deoxyribonucleic acid) (from salmon testes)
Catalog # AAJ60840-03
Supplier: Thermo Scientific Chemicals
CAS Number:
Specifications
- Size:1 g
- Cat. No.:AAJ60840-03
- Supplier no.:J60840-03
Specifications
About this item
:
Storage Temperature:Ambient
CAS Number:9007-49-2
:73049-39-5
MDL Number:MFCD00130922