RIPA buffer-2 (2X), Thermo Scientific
125ml
Supplier: Thermo Scientific Chemicals
: J60629-EQE
RIPA buffer-2 (2X), Thermo Scientific
Catalog # AAJ60629-EQE
Supplier: Thermo Scientific Chemicals
CAS Number:
Jump to:
Product Details & Documents
Liquid
Physical Form: Liquid
Specifications
Specifications
- Size:125ml
- Cat. No.:AAJ60629-EQE
- Physical Form:Liquid
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



